Learn More
Contec™ Drip Caps for Multibucket Systems
Fits Contec™ Bucket Systems. The Contec™ Stainless Steel Drip Cap is easy to install and prevents drips between buckets.
Supplier: Contec™ 2773
Comprend: A stainless-steel connecting kit to prevent drips between the buckets.
Description
- Compatible with all disinfectant solutions and common solvents
- 100% stainless steel for durability and cleanliness
- Autoclavable
Identifiants chimiques
| Autoclavable |
Spécifications
| Autoclavable | |
| 5 gal. multi-Bucket System |