missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Connexin 32 Monoclonal Antibody (CXN-32)
GREENER_CHOICE

Numéro de catalogue. MA125094
Modifier l'affichage
Click to view available options
Quantité:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
MA125094 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. MA125094 Fournisseur Invitrogen™ Code fournisseur MA125094
Saison des économies
Afficher toutes les promotions

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Mouse Monoclonal Antibody

Recommended positive controls: mouse whole brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.

Spécifications

Antigène Connexin 32
Applications ELISA, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone CXN-32
Concentration 9 mg/mL
Conjugué Unconjugated
Formule Ascites with 15mM sodium azide
Gène GJB1
Numéro d’acquisition du gène P08033, P08034, P28230
Alias du gène CMTX; CMTX1; Cnx32; connexin 32; connexin-32; CX32; Cxn-32; GAP junction 28 kDa liver protein; gap junction beta-1 protein; gap junction membrane channel protein beta 1; gap junction protein beta 1; gap junction protein, beta 1; gap junction protein, beta 1, 32kDa; Gjb1
Symboles de gène GJB1
Espèces hôtes Mouse
Immunogène Synthetic connexin-32 peptide (amino acids 105-123).
Quantité 50 μL
Statut réglementaire RUO
Primaire ou secondaire Primary
ID du gène (Entrez) 14618, 2705, 29584
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG1
Afficher plus de résultats Afficher moins de résultats