missing translation for 'onlineSavingsMsg'
Learn More

Congo Red Indicator, 0.1% (w/v), pH 3.0 to 5.0 Blue to Red, Certified, LabChem™

Supplier:  LabChem LC133601

Quantity 500 mL

 View more versions of this product

Catalog No. LC133601


Only null left

Description

Description

Specifications

Chemical Identifiers

573-58-0 , 7732-18-5
696.664
11313
disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
C32H22N6Na2O6S2
IQFVPQOLBLOTPF-UHFFFAOYSA-L
CHEBI:34653
C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Congo Red Indicator
573-58-0 , 7732-18-5
C32H22N6Na2O6S2
Soluble in water
IQFVPQOLBLOTPF-UHFFFAOYSA-L
disodium;4-amino-3-[[4-[4-[(1-amino-4-sulfonatonaphthalen-2-yl)diazenyl]phenyl]phenyl]diazenyl]naphthalene-1-sulfonate
11313
696.67
Poly Bottle
Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Red
Liquid
0.1% (w/v), pH 3.0 to 5.0 Blue to Red
99.9, 0.1
C32H22N6O6S2Na2
Passes Test
C1=CC=C2C(=C1)C(=CC(=C2N)N=NC3=CC=C(C=C3)C4=CC=C(C=C4)N=NC5=C(C6=CC=CC=C6C(=C5)S(=O)(=O)[O-])N)S(=O)(=O)[O-].[Na+].[Na+]
696.664
CHEBI:34653
Certified
1g/mL
1g/mL
500 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause cancer.
Suspected of damaging fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
If exposed or concerned: Get medical advice/attention.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions