missing translation for 'onlineSavingsMsg'
Learn More

Conductivity and TDS Standard, Oakton™

Catalog No. 13300119
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13300119 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13300119 Supplier Oakton Instruments Supplier No. 0065389

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for laboratory and field analysis

  • All Oakton Conductivity and TDS Standards are traceable to NIST standard reference materials
  • Solutions help you calibrate conductivity instruments to ensure accuracy
  • Standard values given at 25°C

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.548
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium;chloride
SMILES [Cl-].[K+]

Specifications

Packaging Bottle
Physical Form Liquid
Technique Calibration of Conductivity Instrument
Color Colorless
pH 4.7 to 7.8
Boiling Point 100°C (212°F)
For Use With (Equipment) Conductivity Meter
Solubility Soluble
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Chemical Name or Material Conductivity Standard
Show More Show Less