accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Buffers and Standards
Electrochemistry Standards
Conductivity and TDS Standards
LC187792
Print
Conductivity Standard, 12,856μmho/cm, Certified, LabChem™
Supplier:
LabChem
LC187792
Catalog No.
LC187792
Sign In
or
Register
to check your price and availability.
$120.71
/
Each
Quantity
Only null left
Explore available promotions
Add to cart
Description
Specifications
Product Suggestions
Videos
SDS
Documents
Promotions
Description
Description
Specifications
Specifications
Chemical Name or Material
Conductivity Standard
Packaging
Poly Bottle
CAS Max %
99.25,0.75
Density
1g/mL
Linear Formula
KCl
Solubility
Soluble in water
Formula Weight
74.55
Name Note
12,856μmho/cm
CAS
7447-40-7
,
7732-18-5
Identification
Passes Test
Physical Form
Liquid
Color
Colorless
Quantity
1 L
Grade
Certified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions