missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 10 μS/cm at 25°C (4.7 ppm TDS as NaCl), Ricca Chemical

Catalog No. 22364
Change view
Click to view available options
Quantity:
120 mL
6 x 20 mL
1 L
20 x 20 mL
4 L
Packaging:
Natural Poly Bottle
Pack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
22364 120 mL Natural Poly Bottle
2236100P 6 x 20 mL Pack
223620 20 x 20 mL Pack
223632 1 L Natural Poly Bottle
22361 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22364 Supplier Ricca Chemical Company Supplier No. 22364
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
Name Note 10μS/cm (10μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 5.0E-4
CAS Max % 100.0, 5.0E-4
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point 0°C
Boiling Point 100°C
Solubility Completely miscible
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 120 mL
Concentration or Composition (by Analyte or Components) 4.7 ppm as NaCl
Show More Show Less