missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Conductivity Meter SevenDirect SD30

Catalog No. 01911922
Change view
Click to view available options
Type:
SevenDirect SD30 Kit
SevenDirect SD30 Pure H2O Kit
SevenDirect SD30, Meter Only
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01911922 SevenDirect SD30 Pure H2O Kit
01911910 SevenDirect SD30, Meter Only
01911921 SevenDirect SD30 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01911922 Supplier Mettler Toledo™ Supplier No. 30671563
Earn Up to 5X the Fisher Scientific Buck$ with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The SevenDirect SD30 is an accurate and flexible benchtop meter for the determination of conductivity, total dissolved solids and salinity. The instrument is the replacement of the SevenCompact and is delivered with an EasyPlace sensor arm.

SevenDirect instruments are benchtop meters developed for a wide range of laboratory applications, which provide accurate measurements of conductivity of almost any sample. The SevenDirect is the successor of the SevenCompact and a great addition to any lab.

  • Large color touchscreen provides direct measurement and easy data entry.
  • Instructions guide users through the sensor calibration process.
  • View detailed measurement information on one measurement screen - or focus on the result only and maximize readability with the uFocus mode.
  • Closed connector compartment and replaceable cover keep the instrument safe from dust and spills, even with sensors attached (IP54 in-use).
  • The instrument stores data like timestamp, sample and sensor ID with every measurement and prints or exports records automatically.
  • A two-level user management protects the most important settings.
  • The EasyPlace electrode arm makes placing the sensor in the sample or buffer container easy.
  • Dedicated positions for cleaning and storage support correct sensor handling.

Specifications

Accuracy (Conductivity) 0.005
Measurement Modes Conductivity
Range (Conductivity) 0.001 μS/cm to 2000 mS/cm
Resolution (Conductivity) 0.001 to 1
Range (Salinity) 0 to 80 ppt
Range (TDS) 0 g/L to 100 g/L
Temperature Range (Metric) -30°C to +130°C
Temperature Range (English) -22°F to +266°F
Resolution (Temperature) 0.1°C
Accuracy (Temperature) ±0.1°C between 0°C and 100°C
Range (Resistivity) 0 to 100 Mohm-cm
Display Type Color TFT
Memory 2000 Standard Measurements
Data Points 2000
Data Management Details PC, Printer, USB Stick
IP Rating IP54
Dimensions (L x W x H) 195 x 205 x 65 mm
Weight (Metric) 850 g
Weight (English) 1.9 lb.
Sensor Types InLab 741-ISM Conductivity/Temperature Sensor
Channels 1
Type SevenDirect SD30 Pure H2O Kit
Show More Show Less