Learn More
Chemglass Inc Condenser, Dewar Type, 24/40 Inner Joint, 350 x 75mm
Numéro de catalogue. CG120905
Encompass
Condenser, Dewar Type, 24/40 Inner Joint, 350 x 75mm. Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen. Condenser has a lower inner drip tip joint and hose connections with an O.D. of 10mm at the middle serration. NOTE: See CG-4519 For Solvent Recovery Traps used with Rotary Evaporators.
Numéro de catalogue. CG120905 Fournisseur Chemglass Inc Code fournisseur CG120905
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Condenser, Dewar Type, 24/40 Inner Joint, 350 x 75mm. Condenser has a large opening at the top for use with dry ice/acetone or liquid nitrogen. Condenser has a lower inner drip tip joint and hose connections with an O.D. of 10mm at the middle serration. NOTE: See CG-4519 For Solvent Recovery Traps used with Rotary Evaporators.