missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Concanavalin A (Con A) Conjugates
Description
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor™ and Alexa Fluor™ Plus conjugates. Concanavalin A (Con A) has been used in a variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC) and flow cytometry (FC). In many cell types, concanavalin A (Con A) is a good marker of ER and Golgi.
Thermo Fisher Scientific offers a broad selection of Invitrogen concanavalin A (Con A) conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Concanavalin A selectively binds to alpha‐mannopyranosyl and alpha‐glucopyranosyl residues. In neutral and alkaline solutions, concanavalin A exists as a tetramer with a molecular weight of approximately 104,000 Daltons. In acidic solutions (pH below 5.0), concanavalin A exists as a dimer. Conjugated concanavalin A has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).
Spécifications
Spécifications
| À utiliser avec (application) | Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
| Quantité | 5 mg |
| Sources | Canavlia ensiformis |
| Conjugué | Alexa Fluor 594 |
| Réactivité croisée | α-man, α-glc |
| Espèces | C. ensiformis |
| Recombinant | Native |
| Étiquette de la protéine | None |
| Contenu et stockage | Store in freezer (-5 to -30°C) and protect from light. |
| Excitation/Émission | 590/617 nm |
| Afficher plus de résultats |