missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Con A Sepharose™ 4B Medium

Catalog No. 45000069
Change view
Click to view available options
Quantity:
5 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000069 5 mL
45000068 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45000069 Supplier Cytiva Supplier No. 17044003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An affinity chromatography media for capture of glycosylated biomolecules including glycoproteins and glycolipids

Highly suited for isolation of cell surface glycoproteins from detergent-solubilized membranes.

  • Con A coupled to Sepharose, in a 4% agarose matrix via cyanogen bromide activation
  • Binds molecules that contain α-D-mannose, α-D-glucose, and sterically related residues with available C-3, C-4, or C-5 hydroxyl groups
  • Group-specific adsorbent for molecules containing sugars
  • Concentration of immobilized Con A, approx. 13mg/mL medium; binding capacity for porcine thyroglobulin, approx. 30mg/mL medium
  • Stable in the pH range 4 to 9, and in the presence of all commonly used buffers
  • Average particle size: 90μm
  • Delivered in 0.1M acetate buffer, pH 6.0, containing 1M NaCl, 1mM CaCl2, 1mM MgCl2, 1mM MnCl2, and 20% ethanol

Specifications

Binding Capacity 30mg/mL
Ligand Concentration 13mg Con A/mL Drained Medium
Matrix 4% Agarose
Particle Size 90 μm
pH 4 to 9
Quantity 5 mL
Content And Storage 2°C to 8°C
For Use With (Application) For separation and purification of Glycoproteins, Polysaccharides and Glycolipids, isolates cell surface Glycoproteins from detergent-solubilized membranes
Ligand Concanavalin A