Learn More
Chemglass Inc Complete Package, 10-20mL Microwave Vials, 28mm OD X 83mm Long
Supplier:  Chemglass Inc CG492002
Encompass
Complete Package, 10-20mL Microwave Vials, 28mm OD X 83mm Long. Perform microwave-heated reactions in glass vials secured by 20mm aluminum seals with 0.125-inch thick, blue PTFE / white silicone septa. Glass vials are made from heavy-wall, Type I, Class A borosilicate for its high resistance to both chemicals and thermal shock. Microwave reaction vials are designed to withstand pressures up to 300 PSI (20 bars). PTFE-coated stir bars are used to provide continuous stirring and even heating during reactions.Chemglass microwave vials come complete with vials, septa and stir bars packed for convenience, cleanliness and protection in white corrugated cartons with removable trays of ten vials each. For crimper and decapper tools see CG-4930.PLEASE NOTE: Stir bars and aluminum seals are supplied but are not assembled.CG-4920-02 Complete Package To Include:100ea 10mL Vials, 100ea 20mm Aluminum Seals with PTFE Faced Silicone Septa and 12ea 6mm Diameter X 15mm Long Egg Shaped PTFE Stir Bars
Numéro de catalogue. 50872808
Il en reste null