Learn More
ABS Compact Pharmacy Refrigerator
Catalog No. 361058651
Change view
Click to view available options
Door Style:
Glass Swing
Solid Swing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Style
361058651 Glass Swing
361058638 Solid Swing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 361058651 Supplier ABS Supplier No. PHABTNSF10PG
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Experience the best in vaccine refrigeration, certified to the NSF/ANSI 456 Vaccine Standard.
These cutting-edge pharmacy refrigerators are certified in accordance with the NSF/ANSI 456 Standard for Vaccine Storage.
  • With this certification, units protect pharmaceuticals at optimal temperatures, preventing waste and allowing for peak delivery.
  • These compact glass door refrigerators utilize microprocessor controllers and feature temperature alarms, remote alarm contacts, and probe access ports with included probes.
  • Units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Specifications
Type Pharmacy Refrigerator
Capacity (English) 10.5 cu. ft.
Capacity (Metric) 297 L
Temperature Range 1°C to 10°C
Door Style Glass Swing
No. of Doors 1
Shelves 7
Cabinet Material Scratch-resistant Painted Steel
Exterior Finish Powder-coated Steel
Certifications/Compliance cULus, ETL, ENERGYSTAR
Style Upright
Monitoring Options Microprocessor Temperature Controller
Compressor H-drive
Plug Type NEMA 5-15
Voltage 115 V
Show More Show Less