missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Compact Digital Dry Bath/Block Heater

Catalog No. 88871001
Change view
Click to view available options
Model:
Compact Dry Bath D
Compact Dry Bath S
Plug Type:
US
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Plug Type
88871001 Compact Dry Bath S US
88871002 Compact Dry Bath D US
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 88871001 Supplier Thermo Scientific™ Supplier No. 88871001

Maximize bench space with our compact, lightweight dry bath with multiple combinations of block sizes for versatility.

Maximize valuable bench space with Thermo Scientific™ Compact Digital Dry Bath/ Block Heater—available with multiple combinations of block sizes for tube size versatility.

  • Built-in temperature-sensing probe assists with outstanding temperature accuracy and control
  • Designed for precise temperature control
  • Fast heating rate
  • Multiple combinations of different sizes of blocks allows for maximum tube size versatility during experimental procedure
  • 12V low voltage power input
  • Two sizes to meet your application volume demands
  • Convenient and easy-to-read digital display
  • Small footprint
  • Lightweight design
  • Single (S) model can hold a full block, 2 half blocks, 4 quarter blocks or 1 half and 2 quarter blocks
  • Double (D) model can hold 2 full blocks, 4 half blocks, 8 quarter blocks or a combination of full/half and quarter blocks.

Applications:

  • Sample preparation
  • Enzyme preservation
  • Enzyme-substrate reactions
  • DNA amplification

Required Accessory:

  • Unit must be ordered with block(s)

Warranty

1 Year, Replacement

Specifications

Certifications/Compliance CE, cTUVus, RoHS
Controller Type Digital
Capacity (Metric) 0.2 mL
Lighting LED
Plug Type US
Temperature Range (Metric) Ambient +2° to 100°C
Type Dry Block Heater
Heating Rate 20 min. average heating rate (30° to 100°C, ambient at 25°C)
Includes Dry Bath, US style plug, block lifter, plastic lid
Model Compact Dry Bath S
No. of Blocks Full block, 2 half blocks, 4 quarter blocks or 1 half and 2 quarter blocks
Temperature Accuracy ≤ ± 0.5°C
Temperature Uniformity ≤ ± 0.5°C
Warranty 1 Year, Replacement
Dimensions (L x W x H) 185 x 185 x 25 mm (7.3 x 7.3 x 1 in.)
Length (English) 7.3 in.
Length (Metric) 185 mm
Width (English) 7.3 in.
Width (Metric) 185 mm
Height (English) 1 in.
Height (Metric) 25 mm
Weight (English) 2.2 lb.
Weight (Metric) <1 kg
Electrical Requirements 12 V, DC, 5 A
Voltage 100/240 V
Show More Show Less

Warranty: 1-year Thermo Fisher Scientific standard, limited warranty. Contact your sales representative for details. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.