missing translation for 'onlineSavingsMsg'
Learn More

Combination Semi-Volatile Surrogate Standard, SPEX CertiPrep™

Catalog No. CLPSSURR
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLPSSURR 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLPSSURR Supplier SPEX CertiPrep Group LLC Supplier No. CLPSSURR

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
2,4,6-Tribromophenol118-79-62000μg/mL
2-Fluorophenol367-12-42000μg/mL
Phenol-d613127-88-32000μg/mL
2-Fluorobiphenyl321-60-81000μg/mL
Nitrobenzene-d54165-60-01000μg/mL
p-Terphenyl-d141718-51-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Combination Semivolatile Surrogate Standard
Packaging 2mL amber prescored glass ampule
Name Note CLP SOW 2/88
Concentration or Composition Notes Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
Methods For US EPA CLP Methods
Technique GC-MS
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix MeCl:Acetone
Quantity 1 mL

Product intended for laboratory use only