Learn More
Morphix Technologies Color Comparators for SafeAir™ Detection Badges
Use to determine the exact exposure dose indicated on a SafeAir badge.
$144.66
Specifications
| For Use With (Application) | Used to determine the exact exposure dose indicated on a SafeAir badge |
|---|
| Catalog Number | Mfr. No. | Detectable Analytes | Measurement Range | Product Type | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Detectable Analytes | Measurement Range | Product Type | Price | Quantity & Availability | |||
189991391
|
Morphix Technologies
383001 |
Hydrazine | 4.5 to 300 ppb/hour (billions) | Hydrazine Detection Badge |
Each for $144.66
|
|
|||
189991392
|
Morphix Technologies
383016 |
Phosgene | 0.9 to 100 ppm.Hour | Phosgene Detection Badge |
Each for $144.66
|
|
|||
Description
- Simple color change technology
- Minimal training
- Rugged
Order Info
Available for hydrazine and phosgene.
Specifications
| Used to determine the exact exposure dose indicated on a SafeAir badge |