Learn More
Morphix Technologies Color Comparators for SafeAir™ Detection Badges
Catalog No. 189991392
Encompass
Change view
Click to view available options
Detectable Analytes:
Hydrazine
Phosgene
Measurement Range:
0.9 to 100 ppm.Hour
4.5 to 300 ppb/hour (billions)
Product Type:
Hydrazine Detection Badge
Phosgene Detection Badge
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes Measurement Range Product Type
189991392 Phosgene 0.9 to 100 ppm.Hour Phosgene Detection Badge
189991391 Hydrazine 4.5 to 300 ppb/hour (billions) Hydrazine Detection Badge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 189991392 Supplier Morphix Technologies Supplier No. 383016
Use to determine the exact exposure dose indicated on a SafeAir badge.
  • Simple color change technology
  • Minimal training
  • Rugged
Order Info
Available for hydrazine and phosgene.
Specifications
For Use With (Application) Used to determine the exact exposure dose indicated on a SafeAir badge
Detectable Analytes Phosgene
Measurement Range 0.9 to 100 ppm.Hour
Product Type Phosgene Detection Badge