Learn More
Morphix Technologies Color Comparators for SafeAir™ Detection Badges
Description
- Simple color change technology
- Minimal training
- Rugged
Order Info
Available for hydrazine and phosgene.
Specifications
Specifications
| For Use With (Application) | Used to determine the exact exposure dose indicated on a SafeAir badge |
| Detectable Analytes | Hydrazine |
| Measurement Range | 4.5 to 300 ppb/hour (billions) |
| Product Type | Hydrazine Detection Badge |