missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collodion, USP, Spectrum™ Chemical
Description
Spectrum™ Chemical Collodion, USP is used as a surgical dressing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 200 L |
| Packaging | Drum |
| UN Number | UN2059 |
| Percent Purity | 5% |
| Grade | USP |