missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit
Description
Features of the TaqMan SARS-CoV-2, FluA/B, RSV RT-PCR Assay Kit include:
- Single assay for SARS-CoV-2, influenza A/B, and RSV RNA—detect and differentiate between infection states and get the information needed to help analyze distribution of SARS-CoV-2, flu, and RSV nucleic acid
- Affordable and scalable—add to your existing SARS-COV-2 assay menu and infrastructure to expand your respiratory sample analysis while maintaining low operational costs and workflow simplicity
- Sensitive and specific—get 100% homology to >99.9% of known SARS‐CoV‐2 genomes* and robust performance for detecting flu A/B and RSV RNA, providing increased confidence in results
- Complete solution for labs—Applied Biosystems Design & Analysis Software automatically converts genetic analysis data into a readable report, helping reduce risk of user interpretation error
Other reagents recommended to be used in conjunction with the TaqMan SARS-CoV-2, FluA/B, RSV RT-PCR Assay Kit include:
- TaqMan SARS-CoV-2, Flu A/B, RSV RNA Control
- TaqMan SARS-CoV-2 Control Dilution Buffer
- TaqPath 1-Step RT-qPCR Master Mix
Specifications
Specifications
| Content And Storage | Store at -30°C to -10°C. |
| Detection Method | Primer-probe |
| Product Line | TaqMan |
| Quantity | 1000 reactions |
| Shipping Condition | Dry Ice |
| Software | Applied Biosystems™ QuantStudio™ Design and Analysis Software 2.4.x |
| Target Organism Class | SARS-CoV-2 (S & N genes), Influenza A/B, RSV |
| No. of Reactions | 1000 reactions |
Frequently Asked Questions (FAQs)
What is the concentration of the MS2 phage provided in the TaqMan SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit (Cat. No. A47702)?
Unfortunately, we cannot disclose the concentration of the MS2 phage in the above kit, as this information is proprietary.
For Research Use Only. Not for use in diagnostic procedures.