missing translation for 'onlineSavingsMsg'
Learn More

CoenzymeQ10, ≥97.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013865
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013865 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013865 Supplier Merck Emd Millipore Supplier No. 0738610MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Coenzyme Q10 is the central electron carrier in mitochondrial respiratory chain, originating from food intake as well as from endogenous synthesis. It commonly acts in its reduced form to prevent lipid peroxidation in biological membranes and in serum low-density lipoproteins (LDL). It also tends to protect the mitochondrial membrane proteins and DNA against oxidative damage along with lipid peroxidation. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 303-98-0
Physical Form Powder
Linear Formula C59H90O4
MDL Number MFCD00042919
Melting Point 49°C
Synonym Q-10; Ubiquinone 50; Ubiquinone-10
Type Analytical Standard
Quantity 10 mg
Formula Weight 863.34
Percent Purity ≥97.5% (HPLC)
Grade Analytical Standard
Show More Show Less