Learn More
Biotium Coelenterazine H
Numéro de catalogue. NC0478370
Encompass
Coelenterazine h (2-(4-Dehydroxy) coelenterazine) is a synthetic derivative of coelenterazine. The luminescence intensity of its aequorin complex is more than 10 times higher than that of the aequorin formed from native coelenterazine. Also see water soluble Aquaphile Coelenterazine h.Properties: Yellow solid soluble in MeOH or EtOH; Store at -20C and protect from light; C26H21N3O2; MW: 407; CAS No. [50909-86-9].
Numéro de catalogue. NC0478370 Fournisseur Biotium Code fournisseur 101111
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Coelenterazine h (2-(4-Dehydroxy) coelenterazine) is a synthetic derivative of coelenterazine. The luminescence intensity of its aequorin complex is more than 10 times higher than that of the aequorin formed from native coelenterazine. Also see water soluble Aquaphile Coelenterazine h.Properties: Yellow solid soluble in MeOH or EtOH; Store at -20C and protect from light; C26H21N3O2; MW: 407; CAS No. [50909-86-9].