missing translation for 'onlineSavingsMsg'
Learn More

Cobaltous Chloride Hexahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. C371500
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C371500 500 g Glass Bottle
C371100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C371500 Supplier Thermo Fisher Scientific Supplier No. FLC371500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
Synonym Cobalt (II) Chloride
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 86°C
Color Violet
pH 4.9
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range 98.0 to 102.0%
Formula Weight 237.93
Percent Purity 98.0 to 102.0%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure Negligible
Chemical Name or Material Cobaltous Chloride Hexahydrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May cause cancer by inhalation. Harmful if swallowed. Irritating to eyes, respiratory system and skin. May cause allergic respiratory and skin reaction. May impair fertility. Possible risks of irreversible effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA
Health:3 Flammability:0 Instability:0
DOTInformation DOT Class 6.1, : Poison