missing translation for 'onlineSavingsMsg'
Learn More

Cobalt, Oil based standard solution, Specpure™ Co 1000μg/g

Catalog No. AA4714318
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4714318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4714318 Supplier Thermo Scientific Chemicals Supplier No. 04714318

missing translation for 'validMainframeMsg'

CAS: 7697-37-2 | HNO3 | 63.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

Physical Form Liquid
Concentration Co 1000µg/g
Quantity 50 g
Grade Specpure
Chemical Name or Material Cobalt, Oil based standard solution
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only