missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Nitrate, Hexahydrate, Practical, Spectrum™ Chemical

Catalog No. 18609856
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609856 45 kg Amber Glass Bottle
18609852 125 g Amber Glass Bottle
18609853 500 g Amber Glass Bottle
18609854 2.5 kg Amber Glass Bottle
18609855 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18609856 Supplier Spectrum Chemical Mfg Cor Supplier No. C131645KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10026-22-9 | 291.03 g/mol

Chemical Identifiers

CAS 10026-22-9

Specifications

CAS Max % 1
Physical Form Crystals and/or Chunks
Quantity 45 kg
Molecular Weight (g/mol) 291.03
Grade Practical
Packaging Amber Glass Bottle
Chemical Name or Material Cobalt Nitrate, Hexahydrate

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.