missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Nitrate, 8% (w/v), LabChem™

Catalog No. LC132207
Change view
Click to view available options
Quantity:
125 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC132207 125 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC132207 Supplier LabChem Supplier No. LC132207
Only null left

Chemical Identifiers

CAS 10026-22-9 , 7732-18-5
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Pink
CAS Max % 92, 8
Physical Form Liquid
Quantity 125 mL
Linear Formula Co(NO3)2·6H2O
Solubility Information Soluble in water
Formula Weight 291.03
Percent Purity 8% (w/v)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Nitrogen oxides; Cobalt and compounds
Chemical Name or Material Cobalt Nitrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature