missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Chloride Hexahydrate 98.0+%, TCI America™

Catalog No. C2388250G
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2388250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2388250G Supplier TCI America Supplier No. C2388250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Color Red
Melting Point 87°C
UN Number 3077
Formula Weight 129.83
Percent Purity ≥98.0% (T)
Quantity 250 g
Physical Form Crystal-Powder at 20°C
Chemical Name or Material Cobalt(II) Chloride Hexahydrate
EINECSNumber (1)-0207
RTECSNumber GG0200000
TSCA Yes