missing translation for 'onlineSavingsMsg'
Learn More

Cytiva CNBr-Activated Sepharose™ Medium

Catalog No. 45000066
Change view
Click to view available options
Quantity:
15 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000066 15 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45000066 Supplier Cytiva Supplier No. 17043001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use for coupling antibodies or other large proteins containing -NH2 groups to the Sepharose media. CNBr-Activated Sepharose™ Medium is a pre-activated media perfect for custom affinity applications.

  • Allows spontaneous and rapid coupling of proteins without the need for an intermediate spacer arm
  • Multipoint attachment resulting in a chemically stable product

Specifications

Ligand Coupling Method Large Protein
Matrix 4% Agarose
Particle Size 90 μm
pH 2 to 11
Quantity 15 g
Content And Storage 2°C to 8°C
For Use With (Application) For coupling ligands containing primary amino groups