missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CMA

Numéro de catalogue. 667750
Modifier l'affichage
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
667750 50 mg
667710 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 667750 Fournisseur Tocris Bioscience™ Code fournisseur 6677/50
Il en reste null

Murine-selective STING agonist

CMA is a STING agonist; murine-selective (Kd = 3.5 μM). Triggers type I IFN response in murine macrophages, with no effect on human STING signaling.

Spécifications

Chemical Name or Material 9-Oxo-10(9H)-acridineacetic acid
CAS 38609-97-1
Synonym ERIS, hMITA, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, sting, transmembrane protein 173
Quantity 50 mg
Target STING Agonists
Molecular Formula C15H11NO3
Formula Weight 253.25
Purity 98%