missing translation for 'onlineSavingsMsg'
Learn More

Clotrimazole 98.0+%, TCI America™

Catalog No. C28675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28675G 5 g
C286725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28675G Supplier TCI America Supplier No. C28675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23593-75-1
Molecular Formula C22H17ClN2
Molecular Weight (g/mol) 344.842
MDL Number MFCD00057220
InChI Key VNFPBHJOKIVQEB-UHFFFAOYSA-N
Synonym clotrimazole, lotrimin, canesten, mycelex, mycosporin, empecid, clotrimazol, mykosporin, gyne lotrimin, chlotrimazole
PubChem CID 2812
ChEBI CHEBI:3764
IUPAC Name 1-[(2-chlorophenyl)-diphenylmethyl]imidazole
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3Cl)N4C=CN=C4

Specifications

Melting Point 147°C
Color White
Quantity 5 g
Formula Weight 344.84
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Clotrimazole
RTECSNumber NI4377000
TSCA No