missing translation for 'onlineSavingsMsg'
Learn More

Clotrimazole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013633
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013633 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013633 Supplier Merck Emd Millipore Supplier No. PHR10581G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Clotrimazole is an imidazole antifungal agent with relatively low toxicity. It is widely used in dermatology and gynaecology applications. Clotrimazole ointments are extensively utilized for the treatment of a wide range of systemic mycoses including candidiasis. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 23593-75-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C22H17 ClN2
MDL Number MFCD00057220
Synonym 1-(o-Chloro-alpha,alpha-diphenylbenzyl)imidazole; 1-(o-Chlorotrityl)imidazole; 1-[(2-Chlorophenyl)diphenylmethyl]-1 H-imidazole
Type Certified Reference Material
Quantity 1 g
Formula Weight 344.84
Grade Certified Reference Material