missing translation for 'onlineSavingsMsg'
Learn More

Cloquintocet-mexyl 98.0+%, TCI America™

Catalog No. C26821G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26821G Supplier TCI America Supplier No. C26821G
Only null left

Chemical Identifiers

CAS 99607-70-2
Molecular Formula C18H22ClNO3
Molecular Weight (g/mol) 335.83
MDL Number MFCD01632329
InChI Key COYBRKAVBMYYSF-UHFFFAOYNA-N
Synonym cloquintocet-mexyl, 1-methylhexyl 5-chloroquinolin-8-yloxy acetate, cloquintocet-mexyl iso, cloquintocet mexyl, cloquintocet-1-methylhexyl ester, heptan-2-yl 2-5-chloroquinolin-8-yl oxyacetate, heptan-2-yl 2-5-chloroquinolin-8-yl oxy acetate, acetic acid, 5-chloro-8-quinolinyl oxy-, 1-methylhexyl ester, rs-1-methylhexyl 5-chloroquinolin-8-yloxy acetate
PubChem CID 93528
IUPAC Name heptan-2-yl 2-[(5-chloroquinolin-8-yl)oxy]acetate
SMILES CCCCCC(C)OC(=O)COC1=C2N=CC=CC2=C(Cl)C=C1

Specifications

Melting Point 70°C
Quantity 1 g
Formula Weight 335.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cloquintocet-mexyl
RTECSNumber AG1265000
TSCA Yes