missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cloperastine Hydrochloride 97.0+%, TCI America™
Supplier: TCI America C303825G
Specifications
| Cloperastine Hydrochloride | |
| 152°C | |
| C20H25Cl2NO | |
| 25 g | |
| UNPLRYRWJLTVAE-UHFFFAOYNA-N | |
| hydrogen 1-{2-[(4-chlorophenyl)(phenyl)methoxy]ethyl}piperidine chloride | |
| 161104 | |
| ≥97.0% (HPLC,T) |
| 14984-68-0 | |
| White-Yellow | |
| MFCD00079012 | |
| 1-[2-[(4-Chlorophenyl)(phenyl)methoxy]ethyl]piperidine Hydrochloride, 4-Chlorobenzhydryl 2-(1-Piperidyl)ethyl Ether Hydrochloride | |
| [H+].[Cl-].ClC1=CC=C(C=C1)C(OCCN1CCCCC1)C1=CC=CC=C1 | |
| 366.33 | |
| 366.33 | |
| Crystalline Powder |
Chemical Identifiers
| 14984-68-0 | |
| 366.33 | |
| UNPLRYRWJLTVAE-UHFFFAOYNA-N | |
| 161104 | |
| [H+].[Cl-].ClC1=CC=C(C=C1)C(OCCN1CCCCC1)C1=CC=CC=C1 |
| C20H25Cl2NO | |
| MFCD00079012 | |
| 1-[2-[(4-Chlorophenyl)(phenyl)methoxy]ethyl]piperidine Hydrochloride, 4-Chlorobenzhydryl 2-(1-Piperidyl)ethyl Ether Hydrochloride | |
| hydrogen 1-{2-[(4-chlorophenyl)(phenyl)methoxy]ethyl}piperidine chloride |
Safety and Handling
RTECSNumber : TM6491500
TSCA : No