accessibility menu, dialog, popup

UserName

HClClON

CAS RN 14984-68-0

IUPAC Name: hydrogen 1-{2-[(4-chlorophenyl)(phenyl)methoxy]ethyl}piperidine chloride
Synonyms: cloperastine
Molecular Weight (g/mol): 366.33
Molecular Formula: C20H25Cl2NO
InChi Key: UNPLRYRWJLTVAE-UHFFFAOYNA-N
SMILES: [H+].[Cl-].ClC1=CC=C(C=C1)C(OCCN1CCCCC1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results