missing translation for 'onlineSavingsMsg'
Learn More

Cloperastine Hydrochloride 97.0+%, TCI America™

Catalog No. p-7125796
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30385G 5 g
C303825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30385G Supplier TCI America Supplier No. C30385G
Only null left

Chemical Identifiers

CAS 14984-68-0
Molecular Formula C20H25Cl2NO
Molecular Weight (g/mol) 366.33
MDL Number MFCD00079012
InChI Key UNPLRYRWJLTVAE-UHFFFAOYNA-N
Synonym 1-[2-[(4-Chlorophenyl)(phenyl)methoxy]ethyl]piperidine Hydrochloride, 4-Chlorobenzhydryl 2-(1-Piperidyl)ethyl Ether Hydrochloride
PubChem CID 161104
IUPAC Name hydrogen 1-{2-[(4-chlorophenyl)(phenyl)methoxy]ethyl}piperidine chloride
SMILES [H+].[Cl-].ClC1=CC=C(C=C1)C(OCCN1CCCCC1)C1=CC=CC=C1

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 5 g
Formula Weight 366.33
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cloperastine Hydrochloride
RTECSNumber TM6491500
TSCA No