missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Clofazimine, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Melting Point | 219°C |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in toluene,slightly soluble in ether,acetone,ethyl acetate |
| Formula Weight | 473.4 |
| Percent Purity | 98% |
| Chemical Name or Material | Clofazimine |
Sécurité et manipulation
| EINECSNumber | 217-980-2 |
RUO – Research Use Only