missing translation for 'onlineSavingsMsg'
Learn More

Clofarabine 98.0+%, TCI America™

Catalog No. C250020MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C250020MG 20 mg
C2500100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C250020MG Supplier TCI America Supplier No. C250020MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123318-82-1
Molecular Formula C10H12ClN5O3
Molecular Weight (g/mol) 285.69
MDL Number MFCD00871077
InChI Key ADXUXRNLBYKGOA-GPGUAWMRNA-N
Synonym clofarabine, clolar, evoltra, clofarex, cafda, clofarabina, clofarabinum, clofarabine usan, cl-f-ara-a, c1-f-ara-a
PubChem CID 119182
ChEBI CHEBI:681569
IUPAC Name (2R,3R,4R,5R)-5-(6-amino-9H-purin-9-yl)-4-chloro-2-(hydroxymethyl)oxolan-3-ol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3Cl)C2=NC=N1

Specifications

Melting Point 227°C
Color White
Quantity 20 mg
Formula Weight 303.68
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Clofarabine
RTECSNumber UD7473000
TSCA No
Recommended Storage Refrigerator