missing translation for 'onlineSavingsMsg'
Learn More

Carson™ Clip n' Clean™ All-in-One Cleaning Kit

Numéro de catalogue. s98190
Encompass
Change view
Click to view available options
:
Cleaning Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue.
S98190 Cleaning Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. S98190 Fournisseur Carson™ Code fournisseur MF50AS
Il en reste null

Designed for people on the go, this all-in-one screen and lens cleaning kit includes cleaning spray, a microfiber cloth, and a protective carrying case.

  • Clip n’ Clean™ base is a small spray bottle that contains an alcohol-free and ammonia-free cleaning solution
  • Kit also includes a double-sided microfiber cloth that will have screens and lenses sparkling in seconds
  • Textured side of the cloth wipes away dust and debris, while the smooth side polishes and shines
  • All items nest securely in the sturdy, lightweight Clip n’ Clean case
  • Conveniently clip onto carabiners, lanyards, and keyrings
  • Small enough to be carried in a purse, pocket, or backpack
  • Perfect for cleaning smartphones, eyeglasses, or sunglasses

Spécifications

Product Type Cleaning Kit
Description Sold in a 4 Pack: Gray (2), Blue (1), Orange (1)
Format All-in-One
For Use With (Application) Cleaning Glasses, Lenses, and Screens
Includes Alcohol-Free/Ammonia-Free Cleaner, Microfiber Cloth, and Protective Case
Length (English) 1.2 in.