missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ ClearVue™ Mountants
Spécifications
Spécifications
| Color | Light yellow |
| Product Type | Mountant |
| Quantity | 1 pt. |
| Solution Type | Mountant, Xylene-based |
| Formulation | Xylenes (o-, m-, p- isomers), Acrylic Resin |
| Form | Liquid |