missing translation for 'onlineSavingsMsg'
Learn More

Mopec ClearSteel, Stainless Steel Cleaning and Polishing Wipes

Catalog No. BE039
Change view
Click to view available options
Product Type:
Wipes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
BE039 Wipes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BE039 Supplier Mopec Supplier No. BE039

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mopec ClearSteel, Stainless Steel Cleaning and Polishing Wipes allow users to clean soiled area while bringing out the natural luster in the equipment and leaving a protective covering that repels water, fingerprints and dust.

  • Specifically designed to be used on all equipment such as workstations, cabinets, coolers and storage areas
  • Wipe in the direction of the stainless steel grain for best results

Specifications

Product Type Wipes
pH 8 to 9
Appearance White
Container Type Canister
Content And Storage Store in a cool dry place
Description Cleaning Wipes
For Use With (Application) Cleaning, Polishing