missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Clear Standard Wide Mouth Glass Bottles without Caps
GREENER_CHOICE

Clear, Type III soda-lime glass ideal for general, small to large volume storage

$29.08 - $122.12

Specifications

Color Clear
Material Glass (Type 3)
Closure Included No
View More Specs

Products 4
Catalog Number Capacity (Metric) Price Quantity & Availability  
Catalog Number Capacity (Metric) Price Quantity & Availability  
FB02911942
GREENER_CHOICE
View Documents Promotion Details
120 mL
Case of 24 Each for $29.08
Only null left
 
FB02911943
GREENER_CHOICE
View Documents Promotion Details
240 mL
Case of 24 Each for $61.96
Only null left
 
FB02912114
GREENER_CHOICE
View Documents Promotion Details
950 mL
Case of 12 Each for $94.36
Only null left
 
FB02911871
GREENER_CHOICE
View Documents Promotion Details
473 mL
Case of 24 Each for $122.12
Only null left
 
Description

Description

  • Features a wide mouth opening for easy transfer of contents
  • Applications include pharmaceutical, chemical, environmental, and food packaging and storing
  • Manufactured from clear flint glass
Specifications

Specifications

Clear
Glass (Type 3)
No
Videos
SDS
Documents

Documents