Learn More
Fisherbrand™ Clear French Square Glass Bottles without Caps
Clear USP Type III soda-lime glass ideal for solvent or chemical, sample storage and sample observation
 View more versions of this product
Catalog No. FB02911947
Only null left
Description
Description
  • General use bottles ideal for when content integrity and sample or product visibility is important
  • Clear glass offers minimal chemical interference with contents and is appropriate when visibility is important but protection from UV rays is not necessary
  • Environmentally sensitive bottles help eliminate waste and help to insure product integrity for long term storage
Specifications
Chemical Identifiers
Clear
No
French Square Bottle
Glass (Type 3)
Wide
Specifications
60 mL
Clear
16 mm
240
No
87 mm
28-400 mm
2 oz.
25 mm
Glass (Type 3)
28-400
Wide
French Square Bottle
39 mm
Product Suggestions
Product Suggestions