missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
GREENER_CHOICE

Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.

$636.93 - $4,706.53

Specifications

Format Standard
Detection Method Colorimetric
Array 8 x 12
Color Clear
Product Type Microplate
View More Specs

Alert:

Each lot is functionally tested for IgG binding.


Products 12
Catalog Number Mfr. No. Surface Treatment Certified Volume (Metric) Working Barcode Price Quantity & Availability  
Catalog Number Mfr. No. Surface Treatment Certified Volume (Metric) Working Barcode Price Quantity & Availability  
1424578
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
N3355
Immulon 1 B No 230 μL No Barcode
Case of 50 Each for $636.93
Only null left
 
1424561
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3455
Immulon 2 HB No 280 μL No Barcode
Case of 50 Each for $704.90
Only null left
 
14245153
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
3855
Immulon 4 HBX No 280 μL No Barcode
Case of 50 Each for $766.43
Only null left
 
12565135
GREENER_CHOICE
Accessories View Documents Promotion Details
Thermo Scientific™
439454
MaxiSorp Yes 350 μL No Barcode
Case of 60 Each for $1,112.48
Only null left
 
12565136
GREENER_CHOICE
Accessories View Documents Promotion Details
Thermo Scientific™
N442404
MaxiSorp No 350 μL No Barcode
Case of 60 Each for $1,050.57
Only null left
 
12565376
View Documents Promotion Details
Thermo Scientific™
456529
PolySorp No 350 μL No Barcode
Case of 180 Each for $3,827.78
Only null left
 
12565377
View Documents Promotion Details
Thermo Scientific™
456537
MaxiSorp Yes 350 μL No Barcode
Case of 180 Each for $4,706.53
Only null left
 
12565730
View Documents Promotion Details
Thermo Scientific™
467320
MediSorp Yes 350 μL No Barcode
Case of 60 Each for $1,555.03
Only null left
 
1496223
View Documents Promotion Details
Thermo Scientific™
467340
MultiSorp No 350 μL No Barcode
Case of 60 Each for $1,666.09
Only null left
 
12565138
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
475094
PolySorp No 350 μL No Barcode
Case of 60 Each for $1,354.38
Only null left
 
21377269
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
9502227
Universal Binding No 330 μL No Barcode
Case of 50 Each for $795.44
Only null left
 
21377270
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
95029330
Enhanced Binding Yes 330 μL No Barcode
Case of 50 Each for $985.84
Only null left
 
Description

Description

Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates:

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces:

  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.

Ordering Information:

Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate

Specifications

Specifications

Standard
8 x 12
Microplate
96
None
Colorimetric
Clear
Polystyrene
Non-sterile
Videos
SDS
Documents

Documents