missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates

Catalog No. 12565377
Change view
Click to view available options
Surface Treatment:
Enhanced Binding
Immulon 1 B
Immulon 2 HB
Immulon 4 HBX
MaxiSorp
MediSorp
MultiSorp
PolySorp
Universal Binding
Certified:
No
Yes
Volume (Metric) Working:
230 μL
280 μL
330 μL
350 μL
Barcode:
No Barcode
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment Certified Volume (Metric) Working Barcode
12565377 MaxiSorp Yes 350 μL No Barcode
1424578 Immulon 1 B No 230 μL No Barcode
1424561 Immulon 2 HB No 280 μL No Barcode
14245153 Immulon 4 HBX No 280 μL No Barcode
12565135 MaxiSorp Yes 350 μL No Barcode
12565136 MaxiSorp No 350 μL No Barcode
12565376 PolySorp No 350 μL No Barcode
12565730 MediSorp Yes 350 μL No Barcode
1496223 MultiSorp No 350 μL No Barcode
12565138 PolySorp No 350 μL No Barcode
21377269 Universal Binding No 330 μL No Barcode
21377270 Enhanced Binding Yes 330 μL No Barcode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Each lot is functionally tested for IgG binding.
Catalog No. 12565377 Supplier Thermo Scientific™ Supplier No. 456537

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Each lot is functionally tested for IgG binding.

Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.

Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

96-Well Solid Plates:

  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces:

  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.

Ordering Information:

Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate

Specifications

Barcode No Barcode
Binding Property Hydrophilic
Certified Yes
Format Standard
Lid Without Lid
Detection Method Colorimetric
Array 8 x 12
Packaging 180 Cases
Binding Type Hydrophilic
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 350 μL
Sterility Non-sterile
Surface Treatment MaxiSorp
Well Shape Round, Flat Bottom (F-well)
No. per Case 180
No. per Pack 10
Plate Blocking None
Quantity 180/Cs.
Target Molecule Antibodies (Glycoproteins)
Show More Show Less