Learn More
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates

Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.
Supplier: Thermo Scientific™ 475094
Alert:
Each lot is functionally tested for IgG binding.
Description
Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
- Clear polystyrene
- Choice of four surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces:
- Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
- Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
- Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
- Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate
Chemical Identifiers
| Standard | |
| 8 x 12 | |
| Microplate | |
| 96 | |
| None |
| Colorimetric | |
| Clear | |
| Polystyrene | |
| Non-sterile |
Specifications
| No Barcode | |
| No | |
| Without Lid | |
| 8 x 12 | |
| Clear | |
| Polystyrene | |
| 96 | |
| Non-sterile | |
| Round, Flat Bottom (F-well) | |
| 5 | |
| 60/Cs. |
| Hydrophobic | |
| Standard | |
| Colorimetric | |
| 60 Cases | |
| Microplate | |
| 400 μL | |
| 350 μL | |
| PolySorp | |
| 60 | |
| None | |
| Lipids |