Learn More
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.
Supplier:  Thermo Scientific™ 467320
 View more versions of this product
Catalog No. 12565730
Only null left
Alert:
Each lot is functionally tested for IgG binding.
Description
Description
Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns
Surfaces:
  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate
Specifications
Chemical Identifiers
Standard
8 x 12
Microplate
96
None
Colorimetric
Clear
Polystyrene
Non-sterile
Specifications
No Barcode
Yes
Without Lid
8 x 12
Clear
Polystyrene
96
Non-sterile
Flat Bottom (F-well)
5
60/Cs.
Slightly Hydrophilic
Standard
Colorimetric
60 Cases
Microplate
400 μL
350 μL
MediSorp
60
None
Proteins
Product Suggestions
Product Suggestions