missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 384-Well Plates
GREENER_CHOICE

Numéro de catalogue. p-4555264
Change view
Click to view available options
Surface Treatment:
Immulon 4 HBX
MaxiSorp
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Surface Treatment
12565347 MaxiSorp
21377838 Immulon 4 HBX
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Each lot is functionally tested for IgG binding.
Numéro de catalogue. 12565347 Fournisseur Thermo Scientific™ Code fournisseur 464718
Il en reste null
Each lot is functionally tested for IgG binding.

Carry out colormetric assays with reliable and reproducible results using these clear, flat-bottom, immuno nonsterile 384-well plates, offered with a choice of surface.

Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 384-Well Plates, offered with a choice of surface for optimal binding. Includes Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.

384-Well Solid Plates:

  • Clear polystyrene
  • Choice of surfaces for optimal binding
  • Alphanumerically labeled rows and columns

Surfaces:

  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo ScientificMaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.

Ordering Information:

Please note that availability may vary by country.

Spécifications

Barcode No Barcode
Binding Property Hydrophilic
Format Standard
Lid Without Lid
Detection Method Colorimetric
Array 16 x 24
Packaging 30 Cases
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 100 μL
Sterility Non-sterile
Surface Treatment MaxiSorp
Well Shape Square, Flat Bottom (F-well)
No. per Case 30
No. per Pack 10
Plate Blocking None
Target Molecule Antibodies (Glycoproteins)
Well Design F384
Afficher plus de résultats Afficher moins de résultats