Learn More
Berkshire™ CleanPath™ Adhesive Mats
Berkshire CleanPath™ Adhesive Mats are the number one choice in cleanroom mats. Place CleanPath™ Adhesive Mats in cleanroom entry areas and change room entry areas to significantly decrease the transfer of foot-borne and wheel-borne contaminants including viable and non-viable particles.
$129.19 - $272.97
Specifications
Color Blue
Material LDPE
Type Adhesive Floor Mat
Quantity 30 Sheets/Mat, 4 Mats/Cs.
View More Specs
Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19066721
View Documents Promotion Details
Berkshire™
CPM18364
36 x 18 in.
Case of 120 Each for $129.19
Only null left
19066722
View Documents Promotion Details
Berkshire™
CPM26454
45 x 26 in.
Case of 120 Each for $168.38
Only null left
19066723
View Documents Promotion Details
Berkshire™
CPM36454
45 x 36 in.
Case of 120 Each for $272.97
Only null left
Description
Description
  • Excellent adhesive strength and cleanliness
  • Blue color allows differentiation for guarding against migration into critical areas
  • Adhesive coated surface film peels off easily for disposal
Compliance
ISO Class 3-8; Cleanroom Class 1 -100,000; EU Grade B/C/D
Specifications
Specifications
Blue
LDPE
Adhesive Floor Mat
30 Sheets/Mat, 4 Mats/Cs.