Learn More
Berkshire™ CleanPath™ Adhesive Mats
Berkshire CleanPath™ Adhesive Mats are the number one choice in cleanroom mats. Place CleanPath™ Adhesive Mats in cleanroom entry areas and change room entry areas to significantly decrease the transfer of foot-borne and wheel-borne contaminants including viable and non-viable particles.
Supplier: Berkshire™ CPM26454
Description
- Excellent adhesive strength and cleanliness
- Blue color allows differentiation for guarding against migration into critical areas
- Adhesive coated surface film peels off easily for disposal
Compliance
ISO Class 3-8; Cleanroom Class 1 -100,000; EU Grade B/C/D
Chemical Identifiers
| Blue | |
| LDPE | |
| Adhesive Floor Mat | |
| 30 Sheets/Mat, 4 Mats/Cs. |
Specifications
| LDPE | |
| Blue | |
| Adhesive Floor Mat | |
| 30 Sheets/Mat, 4 Mats/Cs. | |
| 66 cm |
| 45 in. | |
| 114 cm | |
| 26 in. | |
| 45 x 26 in. |