missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Clathrin Heavy Chain Recombinant Rabbit Monoclonal Antibody (23GB3475)

Numéro de catalogue. MA553623
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA553623 20 μL
MA553622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA553623 Fournisseur Invitrogen™ Code fournisseur MA553623
Il en reste null

Rabbit Recombinant Monoclonal Antibody

Clathrin Heavy Chain Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.

Spécifications

Antigen Clathrin Heavy Chain
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3475
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLTC
Gene Accession No. P11442, P53675, Q00610, Q68FD5
Gene Alias 3110065L21Rik; AtCHC2; CHC; CHC17; CHC2; CHC22; clathrin heavy chain; clathrin heavy chain 1; Clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 17; Clathrin heavy chain on chromosome 22; clathrin, heavy chain (Hc); clathrin, heavy chain-like 1; clathrin, heavy polypeptide (Hc); Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; clathrin, heavy polypeptide-like 2; CLH17; CLH-17; CLH22; CLH-22; Cltc; CLTCL; CLTCL1; CLTCL2; CLTD; Hc; KIAA0034; R74732
Gene Symbols CLTC, CLTCL1
Host Species Rabbit
Immunogen A synthetic peptide of human Clathrin heavy chain (204-256AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1213, 54241, 67300, 8218
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Afficher plus de résultats Afficher moins de résultats