missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Class 2 Laser Pointer
Description
Economical laser pointers are useful for many optical experiments.
- Ideal for middle school and high school classrooms
- Lasers operate on a push-button switch, are housed in a rubberized aluminum pen-style case, and include a pocket clip
- Class 2 Red laser pointer has a maximum output of 1 mW and a wavelength of 650 nm (± 10 nm)
- Class 2 Green laser pointer has a maximum output of 1 mW and a wavelength of 532 nm (± 10 nm)
Spécifications
Spécifications
| Product Type | Laser Pointer |
| Diameter (English) | 0.5 in. |
| Diameter (Metric) | 12.7 mm |
| Length (English) | 6 in. |
| Length (Metric) | 15.24 cm |
| Battery Type | AAA |
| Output Power | 1 mW |
| Color | Green |
| Range | Beam: 125 m |